Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapkapk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008551.1 | NP_032577.1 | 9 | 1023 | ||
GenBank mRNA | AK192111.1 | - | 2 | 141 | ||
AK198261.1 | - | 2 | 141 | |||
BC024559.1 | - | 5 | 392 | |||
BC063064.1 | - | 9 | 1023 | 5687007 | ||
X76850.1 | - | 9 | 1002 |