Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010778.3 | NP_034908.1 | 6 | 835 | ||
GenBank mRNA | AB001566.1 | - | 6 | 812 | ||
AB010919.1 | - | 6 | 832 | |||
AB022600.1 | - | 6 | 812 | |||
AK006632.1 | - | 6 | 836 | |||
BC150660.1 | - | 6 | 836 | 9087571 |