Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meis1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193271.1 | NP_001180200.1 | 3 | 1007 | ||
NM_010789.3 | NP_034919.1 | 3 | 1007 | |||
GenBank mRNA | AB221660.1 | - | 2 | 270 | ||
AK132298.1 | - | 3 | 787 | |||
AK134321.1 | - | 3 | 1002 | |||
AK140748.1 | - | 3 | 1011 | |||
AK168684.1 | - | 3 | 999 | |||
BC023689.1 | - | 3 | 1003 | 5344307 | ||
U33629.1 | - | 3 | 718 | |||
U33630.1 | - | 3 | 718 |