Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008593.3 | NP_032619.1 | 1 | 711 | ||
GenBank mRNA | AF023915.1 | - | 1 | 689 | ||
L10407.1 | - | 1 | 202 |