Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meis2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136072.2 | NP_001129544.1 | 4, 5 | 694 | ||
NM_001159567.1 | NP_001153039.1 | 4, 5 | 694 | |||
NM_001159568.1 | NP_001153040.1 | 4, 5 | 694 | |||
NM_001159569.1 | NP_001153041.1 | 4, 5 | 967 | |||
NM_001159570.1 | NP_001153042.1 | 4, 5 | 967 | |||
NM_010825.3 | NP_034955.1 | 4, 5 | 694 | |||
GenBank mRNA | AJ000504.1 | - | 4, 5 | 683 | ||
AJ000505.1 | - | 4, 5 | 683 | |||
AJ000506.1 | - | 4, 5 | 683 | |||
AJ000507.1 | - | 4, 5 | 683 | |||
AK040611.1 | - | 2, 3 | 975 | |||
AK045126.1 | - | 2, 3 | 439 | |||
AK146633.1 | - | 4, 5 | 694 | |||
AK158513.1 | - | 4, 5 | 859 | |||
AK158839.1 | - | 4, 5 | 966 | |||
BC017375.1 | - | 4, 5 | 760 | 4191098 | ||
KM974644.1 | - | 2, 3 | 177 | |||
U57343.1 | - | 731 | ||||
U68383.1 | - | 4, 5 | 653 | |||
U68384.1 | - | 4, 5 | 653 |