Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ott | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011022.1 | NP_035152.1 | 1334 | |||
GenBank mRNA | AK133020.1 | - | 2, 3 | 205 | ||
X96603.1 | - | 2, 3 | 330 | |||
X96606.1 | - | 1334 |
Gene Symbol: Gm15080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198988.1 | NP_001185917.1 | 5, 6 | 1334 |
Gene Symbol: Gm15085 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122734.1 | NP_001116206.1 | 1334 |
Gene Symbol: Gm15093 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099920.1 | NP_001093390.1 | 1328 |
Gene Symbol: Gm15107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC109131.1 | - | 1303 | 40038981 |
Gene Symbol: Gm15114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082966.1 | NP_001076435.1 | 5, 6 | 1280 |