Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023662.3 | NP_076151.2 | 32 | 5568 | ||
GenBank mRNA | AB029291.1 | - | 32 | 5559 | ||
AF352180.1 | - | 31 | 5342 | |||
AK017964.1 | - | 5 | 687 | |||
AK020493.1 | - | 1 | 13 | |||
AK052710.1 | - | 11 | 1437 | |||
BC139033.1 | - | 32 | 5459 | 8862055 |