Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291184.1 | NP_001278113.1 | 8, 9 | 1215 | ||
NM_011048.1 | NP_035178.1 | 8, 9 | 1215 | |||
GenBank mRNA | BC037450.1 | - | 6, 7 | 820 | ||
D50060.1 | - | 8, 9 | 1138 |