Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdha2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008811.2 | NP_032837.1 | 1 | 1602 | ||
GenBank mRNA | AK006356.1 | - | 1 | 1031 | ||
AK076791.1 | - | 1 | 1074 | |||
BC100460.1 | - | 1 | 1022 | 30918688 | ||
M76728.1 | - | 1 | 1599 |