Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080773.2 | NP_001074242.1 | 7 | 881 | ||
NM_001286662.1 | NP_001273591.1 | 7 | 799 | |||
NM_011062.4 | NP_035192.2 | 7 | 881 | |||
GenBank mRNA | AF086625.1 | - | 7 | 847 | ||
AF126294.1 | - | 7 | 822 | |||
AK147149.1 | - | 7 | 873 | |||
AK147835.1 | - | 7 | 856 | |||
AK149162.1 | - | 7 | 815 | |||
AK149289.1 | - | 7 | 839 | |||
AK162677.1 | - | 7 | 841 | |||
AK165564.1 | - | 7 | 818 | |||
AY062008.1 | - | 7 | 777 | |||
BC146001.1 | - | 7 | 818 | 40131304 | ||
BC146003.1 | - | 7 | 818 | 40131306 |