Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006059.3 | NP_006050.3 | 27 | 4530 | ||
XM_011518121.1 | XP_011516423.1 | 27 | 4491 | |||
GenBank mRNA | AB208887.1 | - | 26 | 3846 | ||
AF041835.1 | - | 28 | 4565 | |||
AL133595.1 | - | 5 | 477 |