Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papola | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011112.3 | NP_035242.1 | 4 | 468 | ||
GenBank mRNA | AK028940.1 | - | 4 | 466 | ||
AK090096.1 | - | 4 | 468 | |||
AK132704.1 | - | 4 | 455 | |||
AK134552.1 | - | 4 | 438 | |||
BC060128.1 | - | 4 | 427 | |||
U52197.1 | - | 4 | 351 | |||
U58134.1 | - | 4 | 412 | |||
U58135.1 | - | 3 | 221 |