Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papola | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011112.3 | NP_035242.1 | 7, 8 | 813 | ||
GenBank mRNA | AK028940.1 | - | 7, 8 | 811 | ||
AK090096.1 | - | 7, 8 | 813 | |||
AK132704.1 | - | 7, 8 | 800 | |||
AK134552.1 | - | 7, 8 | 783 | |||
U52197.1 | - | 7, 8 | 696 | |||
U58134.1 | - | 7, 8 | 757 | |||
U58135.1 | - | 6, 7 | 566 |