Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals3bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011150.2 | NP_035280.1 | 6 | 955 | ||
GenBank mRNA | AK079915.1 | - | 6 | 984 | ||
AK137836.1 | - | 6 | 932 | |||
AK150021.1 | - | 6 | 984 | |||
AK152182.1 | - | 6 | 897 | |||
AK153313.1 | - | 6 | 971 | |||
AK153396.1 | - | 6 | 973 | |||
AK155031.1 | - | 6 | 897 | |||
BC090658.1 | - | 6 | 951 | 30666981 | ||
L16894.1 | - | 6 | 962 | |||
X67809.1 | - | 6 | 955 |