Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRISP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190986.1 | NP_001177915.1 | 8 | 812 | ||
NM_006061.2 | NP_006052.2 | 8 | 800 | |||
GenBank mRNA | AK223380.1 | - | 8 | 704 | ||
AK292786.1 | - | 8 | 759 | |||
AK303966.1 | - | 6 | 527 | |||
AK313387.1 | - | 8 | 733 | |||
BC069580.1 | - | 7 | 686 | 7262396 | ||
BC069602.1 | - | 7 | 688 | 7262408 | ||
BC101539.1 | - | 7 | 710 | 8069045 | ||
X94323.1 | - | 8 | 713 | |||
X95240.1 | - | 7 | 688 |