Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013637.4 | NP_038665.1 | 2 | 221 | ||
GenBank mRNA | AK006377.1 | - | 2 | 220 | ||
AK007068.1 | - | 2 | 221 | |||
AK018980.1 | - | 2 | 224 | |||
BC049599.1 | - | 220 | 6773822 | |||
BC059733.1 | - | 2 | 220 | 6774637 | ||
K02926.1 | - | 2 | 206 | |||
M27500.1 | - | 2 | 206 | |||
X14003.1 | - | 2 | 220 |