Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmsb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039392.2 | NP_001034481.1 | 2 | 344 | ||
NM_001190327.1 | NP_001177256.1 | 4 | 355 | |||
NM_025284.4 | NP_079560.1 | 3 | 283 | |||
GenBank mRNA | AK003178.1 | - | 3 | 200 | ||
AK005512.1 | - | 3 | 200 | |||
AK008557.1 | - | 3 | 200 | |||
AK012361.1 | - | 3 | 200 | |||
AK019212.1 | - | 3 | 200 | |||
AK019232.1 | - | 3 | 201 | |||
AK131711.1 | - | 3 | 200 | |||
AK161987.1 | - | 3 | 200 | |||
BC037153.1 | - | 3 | 191 | |||
BC038926.1 | - | 3 | 201 | |||
BC048070.1 | - | 3 | 179 | |||
BC059777.1 | - | 3 | 187 | |||
BC070416.1 | - | 3 | 178 | |||
BC080312.1 | - | 3 | 181 | |||
BC089326.1 | - | 3 | 159 | |||
BC089327.1 | - | 2 | 139 | |||
BC092009.1 | - | 3 | 184 | |||
BC093587.1 | - | 3 | 185 | |||
BC094284.1 | - | 3 | 173 | |||
BC099374.1 | - | 3 | 182 | |||
BC145877.1 | - | 3 | 160 | 40131180 | ||
BC145879.1 | - | 3 | 160 | 40131182 | ||
CA466695.1 | - | 347 | ||||
CB273987.1 | - | 2 | 290 | |||
CJ042349.1 | - | 2 | 414 | |||
CJ042420.1 | - | 2 | 286 | |||
Z48496.1 | - | 2 | 277 |