Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006063.2 | NP_006054.2 | 4, 5 | 1650 | ||
GenBank mRNA | AF056929.1 | - | 4, 5 | 1620 | ||
AF333387.1 | - | 4, 5 | 1650 | |||
AK316571.1 | - | 4, 5 | 1647 | |||
AY364252.1 | - | 4, 5 | 1060 | |||
BC006534.2 | - | 4, 5 | 1628 | 2959774 |