Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm13023 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007077.2 | NP_001007078.2 | 3 | 715 | ||
GenBank mRNA | AK135972.1 | - | 3 | 698 | ||
BC084594.1 | - | 3 | 696 | 30478700 |
Gene Symbol: C87977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177542.1 | NP_001171013.1 | 3 | 699 | ||
GenBank mRNA | AK139502.1 | - | 3 | 698 | ||
BC094895.1 | - | 3 | 671 |
Gene Symbol: Gm10436 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105254.1 | NP_001098724.1 | 3 | 772 | ||
GenBank mRNA | AK139576.1 | - | 2 | 357 | ||
BC094894.1 | - | 2 | 334 |
Gene Symbol: Gm2042 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270792.1 | NP_001257721.1 | 3 | 720 | ||
GenBank mRNA | AK135749.1 | - | 3 | 698 | ||
AK139641.1 | - | 3 | 697 | |||
BC086484.1 | - | 3 | 693 | 30476881 |
Gene Symbol: Oog1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178657.5 | NP_848772.3 | 3 | 720 | ||
GenBank mRNA | AB050008.3 | - | 1 | 310 | ||
AB077322.2 | - | 1 | 310 | |||
AK145346.1 | - | 3 | 700 |