Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159482.1 | NP_001152954.1 | 11 | 1311 | ||
NM_033475.3 | NP_258436.2 | 10 | 1656 | |||
GenBank mRNA | AB082927.1 | - | 10 | 1569 | ||
AF327929.1 | - | 11 | 1261 | |||
AK160308.1 | - | 1625 | ||||
AK207859.1 | - | 1 | 255 | |||
AK217557.1 | - | 1 | 255 | |||
AV142766.1 | - | 100 | ||||
BC038638.1 | - | 10 | 1131 | 5058535 | ||
S72304.1 | - | 8 | 914 |