Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ran | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009391.3 | NP_033417.1 | 7 | 1225 | ||
GenBank mRNA | AK075900.1 | - | 7 | 1163 | ||
AK151341.1 | - | 7 | 1157 | |||
AK151937.1 | - | 7 | 1157 | |||
AK153411.1 | - | 7 | 1157 |