Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ranbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011239.2 | NP_035369.2 | 2, 3 | 294 | ||
GenBank mRNA | AK002989.1 | - | 2, 3 | 260 | ||
AK008276.1 | - | 2, 3 | 257 | |||
AK088781.1 | - | 2, 3 | 282 | |||
AK151167.1 | - | 2, 3 | 272 | |||
AK153074.1 | - | 2, 3 | 276 | |||
AK166642.1 | - | 2, 3 | 296 | |||
AK167053.1 | - | 2, 3 | 278 | |||
BC061140.1 | - | 2, 3 | 313 | 6775220 | ||
L25255.1 | - | 2, 3 | 167 | |||
X56045.1 | - | 2, 3 | 299 | |||
X56046.1 | - | 2, 3 | 299 |