Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF169300.1 | - | 6 | 579 | ||
AK019171.1 | - | 6 | 594 | |||
AK019656.1 | - | 6 | 630 | |||
AK090162.1 | - | 6 | 617 | |||
AK133924.1 | - | 6 | 679 | |||
AK145596.1 | - | 6 | 556 | |||
AK147057.1 | - | 6 | 539 | |||
AK151116.1 | - | 6 | 587 | |||
AK158987.1 | - | 6 | 596 | |||
AK159296.1 | - | 6 | 594 | |||
AK159778.1 | - | 6 | 588 | |||
AK159949.1 | - | 6 | 636 | |||
BC003282.1 | - | 6 | 558 | 3582133 | ||
U95141.1 | - | 4 | 269 |