Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080127.1 | NP_001073596.1 | 6 | 723 | ||
NM_001080128.1 | NP_001073597.1 | 5 | 542 | |||
NM_009070.2 | NP_033096.2 | 7 | 845 | |||
GenBank mRNA | AK011475.1 | - | 6 | 725 | ||
AK151150.1 | - | 6 | 713 | |||
AK165905.1 | - | 5 | 529 | |||
BC008089.1 | - | 3 | 280 |