Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011288.1 | NP_035418.1 | 5 | 389 | ||
GenBank mRNA | AB049646.1 | - | 5 | 343 | ||
AK086805.1 | - | 5 | 343 | |||
BC038626.1 | - | 5 | 361 | 4008503 | ||
BC081460.1 | - | 5 | 329 | 6814073 | ||
U84902.1 | - | 5 | 331 |