Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRPB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005260641.3 | XP_005260698.1 | 2 | 341 | ||
XM_005260643.3 | XP_005260700.1 | 2 | 341 | |||
XM_005260644.3 | XP_005260701.1 | 2 | 342 | |||
XM_017027577.1 | XP_016883066.1 | 2 | 777 | |||
XM_017027578.1 | XP_016883067.1 | 2 | 782 | |||
XM_017027579.1 | XP_016883068.1 | 2 | 783 | |||
XM_017027580.1 | XP_016883069.1 | 2 | 783 | |||
XM_017027581.1 | XP_016883070.1 | 2 | 780 | |||
XM_017027582.1 | XP_016883071.1 | 2 | 793 | |||
XM_017027583.1 | XP_016883072.1 | 2 | 777 | |||
XM_017027584.1 | XP_016883073.1 | 2 | 777 | |||
XM_017027585.1 | XP_016883074.1 | 2 | 774 | |||
GenBank mRNA | AK313517.1 | - | 2 | 345 | ||
BC025286.1 | - | 2 | 332 | |||
BC075835.1 | - | 2 | 332 | |||
BC131747.1 | - | 2 | 316 | |||
DA331023.1 | - | 2 | 342 | |||
DA967881.1 | - | 2 | 398 | |||
Y10376.1 | - | 2 | 320 |
Gene Symbol: SIRPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DA466740.1 | - | 2 | 164 |