Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009091.2 | NP_033117.1 | 3, 4 | 384 | ||
GenBank mRNA | AA027738.1 | - | 2, 3 | 311 | ||
AK146147.1 | - | 3, 4 | 356 | |||
AV569793.1 | - | 3, 4 | 384 | |||
BC010763.1 | - | 2, 3 | 350 | 4234356 | ||
BC094409.1 | - | 2, 3 | 349 | 1179896 | ||
M33330.1 | - | 3, 4 | 364 |