Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013661.2 | NP_038689.2 | 14, 15 | 2253 | ||
GenBank mRNA | AK078659.1 | - | 2, 3 | 185 | ||
AK129362.1 | - | 4, 5 | 580 | |||
BC052397.1 | - | 14, 15 | 2253 | 5719939 | ||
X97818.1 | - | 14, 15 | 2044 |