Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253804.1 | NP_001240733.1 | 18, 19 | 2516 | ||
NM_009195.3 | NP_033221.1 | 18, 19 | 2510 | |||
GenBank mRNA | AF047339.1 | - | 18, 19 | 2434 | ||
AF121118.1 | - | 18, 19 | 2501 | |||
AK149969.1 | - | 18, 19 | 2499 | |||
AK151717.1 | - | 18, 19 | 2471 | |||
AK152762.1 | - | 18, 19 | 2488 | |||
AK153245.1 | - | 18, 19 | 2488 | |||
AK159274.1 | - | 18, 19 | 2498 | |||
AK159331.1 | - | 18, 19 | 2493 | |||
AK159483.1 | - | 18, 19 | 2466 | |||
AK159537.1 | - | 18, 19 | 2493 | |||
AK160034.1 | - | 18, 19 | 2459 | |||
AK160072.1 | - | 18, 19 | 2493 | |||
AK164479.1 | - | 18, 19 | 2475 | |||
AK165874.1 | - | 18, 19 | 2488 | |||
AK166304.1 | - | 18, 19 | 2469 | |||
AK167371.1 | - | 18, 19 | 2507 | |||
AK167444.1 | - | 18, 19 | 2507 | |||
AK169044.1 | - | 18, 19 | 2449 | |||
AK185898.1 | - | 1, 2 | 19 | |||
AK193687.1 | - | 1, 2 | 19 | |||
AK200839.1 | - | 1, 2 | 19 | |||
BC066872.1 | - | 18, 19 | 2440 | 30436525 |