Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009196.4 | NP_033222.1 | 4 | 868 | ||
GenBank mRNA | AF058055.1 | - | 4 | 849 | ||
AK033516.1 | - | 4 | 866 | |||
AK053421.1 | - | 1 | 229 | |||
AK079379.1 | - | 4 | 864 | |||
AK088762.1 | - | 4 | 866 | |||
AK161701.1 | - | 4 | 866 | |||
AK162682.1 | - | 4 | 865 | |||
AK166606.1 | - | 4 | 866 | |||
BC014777.1 | - | 4 | 876 | 3499773 | ||
X82438.1 | - | 3 | 660 |