Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009215.1 | NP_033241.1 | 2 | 337 | ||
GenBank mRNA | AK003014.1 | - | 2 | 338 | ||
BC010770.1 | - | 2 | 327 | 4218815 |