Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpine2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009255.4 | NP_033281.1 | 8 | 1306 | ||
GenBank mRNA | AK045954.1 | - | 8 | 1278 | ||
AK047856.1 | - | 8 | 1278 | |||
AK051332.1 | - | 8 | 1278 | |||
AK159687.1 | - | 8 | 1278 | |||
BC010675.1 | - | 8 | 1256 | 3595407 | ||
CT010311.1 | - | 7 | 1129 | |||
X70296.1 | - | 8 | 1277 |