Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025287.2 | NP_079563.2 | 8, 9 | 895 | ||
GenBank mRNA | AB071989.1 | - | 6, 7 | 704 | ||
AK004669.1 | - | 8, 9 | 865 | |||
AK028201.1 | - | 7, 8 | 1031 | |||
AK030746.1 | - | 7, 8 | 1082 | |||
AK132383.1 | - | 7, 8 | 1071 | |||
AK159482.1 | - | 7, 8 | 1048 | |||
AK160858.1 | - | 7, 8 | 993 | |||
AK166581.1 | - | 7, 8 | 1005 | |||
AK168185.1 | - | 7, 8 | 1017 | |||
AK168546.1 | - | 7, 8 | 1084 | |||
AK189454.1 | - | 3, 4 | 296 | |||
AK192436.1 | - | 2, 3 | 155 | |||
AK198534.1 | - | 2, 3 | 155 | |||
AK199623.1 | - | 2, 3 | 155 | |||
AY538613.1 | - | 6, 7 | 707 | |||
BC005793.1 | - | 1, 2 | 14 | |||
BC043131.1 | - | 7, 8 | 914 | 6811159 | ||
BC045205.1 | - | 8, 9 | 865 | 4988521 | ||
BC064809.1 | - | 7, 8 | 1019 |