Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278237.1 | NP_001265166.1 | 4, 5 | 909 | ||
NM_014254.2 | NP_055069.1 | 4, 5 | 899 | |||
XM_005268562.3 | XP_005268619.1 | 5, 6 | 1009 | |||
XM_005268563.3 | XP_005268620.1 | 5, 6 | 1082 | |||
XM_017018686.1 | XP_016874175.1 | 5, 6 | 964 | |||
XM_017018687.1 | XP_016874176.1 | 6, 7 | 1126 | |||
GenBank mRNA | AB015633.1 | - | 4, 5 | 845 | ||
AK289382.1 | - | 4, 5 | 793 | |||
BC002596.2 | - | 4, 5 | 830 | |||
BC013152.1 | - | 4, 5 | 831 | 3919181 | ||
BX647105.1 | - | 4, 5 | 898 | |||
BX647419.1 | - | 4, 5 | 898 |