Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stfa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025288.2 | NP_079564.1 | 2, 3 | 209 | ||
GenBank mRNA | AK014349.1 | - | 2, 3 | 212 | ||
AY169236.1 | - | 2, 3 | 192 | |||
BC107224.1 | - | 2, 3 | 203 | 40051891 | ||
BC107225.1 | - | 2, 3 | 205 | 40051892 | ||
M92419.1 | - | 2, 3 | 193 |