Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153056.2 | NP_694696.2 | 7 | 740 | ||
GenBank mRNA | AB221687.1 | - | 3 | 260 | ||
AK135484.1 | - | 6 | 810 | |||
AY251540.1 | - | 7 | 716 | |||
BC026403.1 | - | 7 | 705 | 4209498 | ||
BC026637.1 | - | 6 | 781 | |||
BC026650.1 | - | 7 | 708 | 4987746 | ||
BC040759.1 | - | 7 | 705 | 4209498 | ||
BC080304.1 | - | 6 | 1044 |