Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285893.1 | NP_001272822.1 | 39 | 3720 | ||
NM_001285894.1 | NP_001272823.1 | 39 | 3583 | |||
GenBank mRNA | AK029007.1 | - | 23 | 1937 | ||
AK031205.1 | - | 16 | 1762 | |||
AK122375.1 | - | 39 | 3532 | |||
AK141678.1 | - | 39 | 3643 | |||
BC023914.1 | - | 25 | 2111 | 5323126 | ||
BC032214.1 | - | 19 | 1689 | 5323693 | ||
BC055323.1 | - | 39 | 3637 | 5706288 |