Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GNT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014256.3 | NP_055071.2 | 2 | 398 | ||
XM_011527626.2 | XP_011525928.1 | 2 | 357 | |||
GenBank mRNA | AB015630.1 | - | 2 | 368 | ||
AB049585.1 | - | 2 | 353 | |||
AF293973.1 | - | 1 | 264 | |||
AJ130847.1 | - | 1 | 251 | |||
AJ278961.1 | - | 1 | 251 | |||
AK075434.1 | - | 3 | 371 | |||
AK075466.1 | - | 2 | 373 | |||
AK225421.1 | - | 2 | 365 | |||
AK225478.1 | - | 2 | 344 | |||
AK314323.1 | - | 2 | 369 | |||
AY358955.1 | - | 2 | 353 | |||
BC066876.1 | - | 2 | 370 | 6971715 | ||
BC067423.1 | - | 2 | 349 | 6971716 | ||
DA385410.1 | - | 2 | 398 |