Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1ar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145964.2 | NP_666076.2 | 6 | 571 | ||
GenBank mRNA | AK200436.1 | - | 248 | |||
BB611022.1 | - | 6 | 259 |