Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nhlrc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172501.2 | NP_766089.1 | 4 | 625 | ||
GenBank mRNA | AK029465.1 | - | 4 | 620 | ||
AK033167.1 | - | 4 | 625 | |||
AK133986.1 | - | 575 | ||||
AK220477.1 | - | 542 | ||||
BC087903.1 | - | 4 | 612 | 30250042 | ||
BC107356.1 | - | 4 | 613 | 40055875 |