Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011538.2 | NP_035668.2 | 3 | 536 | ||
GenBank mRNA | AY654733.1 | - | 3 | 512 | ||
BF607954.1 | - | 3 | 571 | |||
U57331.1 | - | 3 | 536 |