Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sidt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289668.1 | NP_001276597.1 | 4 | 826 | ||
NM_172257.4 | NP_758461.1 | 4 | 826 | |||
GenBank mRNA | AK081177.1 | - | 4 | 812 | ||
AK152371.1 | - | 4 | 759 | |||
AK155875.1 | - | 4 | 730 | |||
AK180084.1 | - | 2 | 85 | |||
AK180621.1 | - | 2 | 85 | |||
BC023957.1 | - | 4 | 830 | 5321190 | ||
BC051101.1 | - | 4 | 554 | 5052849 | ||
CK639655.1 | - | 2 | 157 |