Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145420.2 | NP_663395.1 | 4 | 320 | ||
GenBank mRNA | AK043167.1 | - | 3 | 2539 | ||
AK144468.1 | - | 4 | 275 | |||
AK148364.1 | - | 4 | 308 | |||
AK187403.1 | - | 3 | 116 | |||
BC019464.1 | - | 4 | 291 | 4205941 | ||
BY111155.1 | - | 4 | 322 |