Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCGF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317836.1 | NP_001304765.1 | 6 | 690 | ||
NM_006315.5 | NP_006306.2 | 5 | 591 | |||
GenBank mRNA | AK057124.1 | - | 5 | 1528 | ||
AK125801.1 | - | 6 | 1535 | |||
AK225892.1 | - | 5 | 590 | |||
AK300290.1 | - | 6 | 599 | |||
AU132917.1 | - | 5 | 548 | |||
BC107061.2 | - | 3 | 216 | 40006331 | ||
BQ882065.1 | - | 5 | 434 | |||
BX648409.1 | - | 1 | 939 |