Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd300a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170758.3 | NP_739564.1 | 2 | 428 | ||
GenBank mRNA | AB065156.1 | - | 2 | 147 | ||
AB091765.1 | - | 2 | 166 | |||
AB091766.1 | - | 2 | 150 | |||
AK045869.1 | - | 2 | 428 | |||
AK053160.1 | - | 2 | 116 | |||
AY457054.1 | - | 2 | 104 | |||
BC139296.1 | - | 2 | 364 | 8862318 | ||
BC139297.1 | - | 2 | 364 | 8862319 |