Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd300ld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145437.2 | NP_663412.1 | 3 | 475 | ||
GenBank mRNA | AB292062.1 | - | 3 | 399 | ||
AK028662.1 | - | 3 | 487 | |||
AK041292.1 | - | 3 | 511 | |||
AY457051.1 | - | 3 | 399 | |||
BC019814.1 | - | 3 | 475 | 5125383 |