Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfpi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009364.3 | NP_033390.1 | 3 | 433 | ||
GenBank mRNA | AK132268.1 | - | 3 | 455 | ||
BC021639.1 | - | 3 | 468 | 4023742 | ||
D50586.1 | - | 3 | 438 |