Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkx2-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146198.1 | NP_001139670.1 | 2 | 2045 | ||
NM_009385.3 | NP_033411.3 | 3 | 2612 | |||
GenBank mRNA | AK136362.1 | - | 1 | 146 | ||
BC057607.1 | - | 3 | 2615 | 6416507 | ||
BC080868.1 | - | 2 | 2095 | 30363051 |