Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf11d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199015.4 | NP_950180.3 | 4 | 368 | ||
GenBank mRNA | AK156583.1 | - | 4 | 349 | ||
BC030186.1 | - | 4 | 369 | 5067268 | ||
BC110663.1 | - | 4 | 325 | |||
BC145673.1 | - | 4 | 288 | 40130976 | ||
BG083212.2 | - | 4 | 368 |