Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009428.2 | NP_033454.1 | 8 | 2327 | ||
GenBank mRNA | AF029983.1 | - | 8 | 1982 | ||
AF060107.1 | - | 8 | 2326 | |||
AJ006204.1 | - | 7 | 1924 | |||
AK047867.1 | - | 6 | 1348 | |||
BC112972.1 | - | 8 | 2145 | 40049027 |